Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM610

Boron-10B solution

IRMM®, certified reference material, aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product IRMM610 is not currently sold in your country. Contact Technical Service

grade

certified reference material

agency

IRMM®

form

aqueous solution

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM610

Legal Information

IRMM is a registered trademark of European Commission

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service