IRMM528RC
Al alloy
IRMM®, certified reference material, 1.0% Co, 1.0 mm wire
Synonym(s):
Aluminum alloy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
agency
IRMM®
manufacturer/tradename
JRC
application(s)
pharmaceutical (small molecule)
format
matrix material
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Analysis Note
For more information please see:
IRMM528RC
IRMM528RC
Legal Information
IRMM is a registered trademark of European Commission
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service