Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM525B

Niobium

IRMM®, certified reference material, 0.1 mm foil

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

resistivity

13-16 μΩ-cm, 20°C

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

application(s)

industrial hygiene
pharmaceutical (small molecule)

format

matrix material

SMILES string

[Nb]

InChI

1S/Nb

Inchi Key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM525B

Legal Information

IRMM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon-Jun Kim et al.
ACS nano, 7(1), 732-739 (2012-12-25)
Imaging the three-dimensional atomic-scale structure of complex interfaces has been the goal of many recent studies, due to its importance to technologically relevant areas. Combining atom-probe tomography and aberration-corrected scanning transmission electron microscopy (STEM), we present an atomic-scale study of
Evgeniy Papulovskiy et al.
Physical chemistry chemical physics : PCCP, 15(14), 5115-5131 (2013-03-02)
Ab initio DFT calculations of (93)Nb NMR parameters using the NMR-CASTEP code were performed for a series of over fifty individual niobates, and a good agreement has been found with experimental NMR parameters. New experimental and calculated (93)Nb NMR data
M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Kulbir Kaur Ghuman et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085501-085501 (2013-01-25)
A systematic study of electronic structure and band gap states is conducted to analyze the monodoping and charge compensated codoping of rutile TiO(2) with Rh and Nb, using the DFT + U approach. Doping of rutile TiO(2) with Rh atoms induces hybridized
Xi Yang et al.
Optics express, 21(4), 4783-4789 (2013-03-14)
Surface dark solitons in photovoltaic nonlinear media are reported. Taking advantage of diffusion and photovoltaic nonlinearities we demonstrated the surface dark solitons and their behaviors near surface theoretically and experimentally in LiNbO₃ crystal. It is very interesting that surface dark

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service