IRMM501C
238UO2 (V encapsulated) (Nuclear Reference Materials)
IRMM®, certified reference material
Synonym(s):
Uraninite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
UNSPSC Code:
12352200
Recommended Products
grade
certified reference material
agency
IRMM®
mass shift
depleted
SMILES string
O=[238U]=O
InChI
1S/2O.U
Inchi Key
FCTBKIHDJGHPPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
IRMM is a registered trademark of European Commission
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service