Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM468

Thyroxine (T4)

IRMM®, certified reference material

Synonym(s):

L-Thyroxine, 3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Beilstein/REAXYS Number:
2228515
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

mp

223 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1

InChI key

XUIIKFGFIJCVMT-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and, thus, protein synthesis.

Analysis Note

For more information please see:
IRMM468

Legal Information

IRMM is a registered trademark of European Commission

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

target_organs

Thyroid,Cardio-vascular system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linnea R Vose et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(44), 17232-17246 (2013-11-01)
Intraventricular hemorrhage (IVH) remains a major cause of white matter injury in preterm infants with no viable therapeutic strategy to restore myelination. Maturation of oligodendrocytes and myelination is influenced by thyroid hormone (TH) signaling, which is mediated by TH receptor
Michaela Granfors et al.
Obstetrics and gynecology, 124(1), 10-15 (2014-06-06)
To evaluate the efficacy of a targeted thyroid testing approach during pregnancy in clinical practice. This is a retrospective cohort study performed within Uppsala County, Sweden. Data were derived from the population-based Uppsala Biobank of Pregnant Women, in which blood
Peter K Quashie et al.
Journal of virology, 89(6), 3163-3175 (2015-01-02)
Dolutegravir (DTG) is the latest antiretroviral (ARV) approved for the treatment of human immunodeficiency virus (HIV) infection. The G118R substitution, previously identified with MK-2048 and raltegravir, may represent the initial substitution in a dolutegravir resistance pathway. We have found that
B Velkeniers et al.
Human reproduction update, 19(3), 251-258 (2013-01-19)
BACKGROUND Previous meta-analyses of observational data indicate that pregnant women with subclinical hypothyroidism have an increased risk of adverse pregnancy outcome. Potential benefits of levothyroxine (LT4) supplementation remain unclear, and no systematic review or meta-analysis of trial findings is available
Steffen Mayerl et al.
The Journal of clinical investigation, 124(5), 1987-1999 (2014-04-03)
Allan-Herndon-Dudley syndrome (AHDS), a severe form of psychomotor retardation with abnormal thyroid hormone (TH) parameters, is linked to mutations in the TH-specific monocarboxylate transporter MCT8. In mice, deletion of Mct8 (Mct8 KO) faithfully replicates AHDS-associated endocrine abnormalities; however, unlike patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service