Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM3702

Zinc 64Zn solution (certified for isotope abundance ratio)

IRMM®, certified reference material, 1 M in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

concentration

1 M in nitric acid

application(s)

pharmaceutical (small molecule)

format

single component solution

mass shift

depleted

SMILES string

[64Zn]

InChI

1S/Zn/i1-1

InChI key

HCHKCACWOHOZIP-BJUDXGSMSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM3702

Legal Information

IRMM is a registered trademark of European Commission

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service