Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM355

Enteroccocus faecalis (CIP 106877)

IRMM®, certified reference material

Synonym(s):

Enteroccocus faecalis in material spheres

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

storage temp.

−70°C

Application

measurements of enterococcus faecalis by colony counting on horse blood agar or Slanetz and Bartley agar according to ISO 7218 and ISO 7899-2 respectively

Analysis Note

For more information please see:
IRMM355

Legal Information

IRMM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service