Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM016

Lithium carbonate-7Li

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7Li2CO3
Molecular Weight:
74.04
UNSPSC Code:
41116107

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

application(s)

pharmaceutical (small molecule)

format

neat

General description

Isotope abundance ratio

Application

Lithium carbonate-7Li has been used as an internal standard to rectify mass discrimination in the boron isotope analysis v using inductively coupled plasma mass spectrometry (ICP-MS) or thermal ionization mass spectrometry.[1]

Analysis Note

For more information please see:
IRMM016

Legal Information

IRMM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of microsublimation and ion exchange chromatography for boron isolation preceding its isotopic analysis via multi-collector ICP-MS
Hoecke V.K, et al.
Journal of Analytical Atomic Spectrometry, 29(10), 1819-1826 (2014)
Anne-Sophie Hanak et al.
Toxicological sciences : an official journal of the Society of Toxicology, 143(1), 185-195 (2014-10-31)
Lithium-induced neurotoxicity may be life threatening. Three patterns have been described, including acute, acute-on-chronic, and chronic poisoning, with unexplained discrepancies in the relationship between clinical features and plasma lithium concentrations. Our objective was to investigate differences in plasma, erythrocyte, cerebrospinal
Anna Thorfve et al.
PloS one, 9(7), e102597-e102597 (2014-07-23)
Bone development and regeneration is associated with the Wnt signaling pathway that, according to literature, can be modulated by lithium ions (Li+). The aim of this study was to evaluate the gene expression profile during peri-implant healing of poly(lactic-co-glycolic acid)
Nicoletta Croce et al.
Journal of psychopharmacology (Oxford, England), 28(10), 964-972 (2014-04-05)
One of the common effects of lithium (Li) and valproic acid (VPA) is their ability to protect against excitotoxic insults. Neurodegenerative and neuropsychiatric diseases may be also associated with altered trophic support of brain-derived neurotrophic factor (BDNF), the most widely
Kazumasu Sasaki et al.
British journal of pharmacology, 172(2), 654-667 (2014-06-11)
μ Opioid receptor knockout (MOP-KO) mice display several behavioural differences from wild-type (WT) littermates including differential responses to nociceptive stimuli. Brain structural changes have been tied to behavioural alterations noted in transgenic mice with targeting of different genes. Hence, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service