Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM011

Boric acid (isotope ratio)

IRMM®, certified reference material, solid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

agency

IRMM®

form

solid

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

10-30°C

Analysis Note

For more information please see:
IRMM011

Legal Information

IRMM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service