Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2637

Supelco

Pasiniazid

analytical standard

Synonym(s):

Isonicotinic acid hydrazide p-aminosalicylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O · C7H7NO3
CAS Number:
Molecular Weight:
290.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

NNC(=O)c1ccncc1.Nc2ccc(C(O)=O)c(O)c2

InChI

1S/C7H7NO3.C6H7N3O/c8-4-1-2-5(7(10)11)6(9)3-4;7-9-6(10)5-1-3-8-4-2-5/h1-3,9H,8H2,(H,10,11);1-4H,7H2,(H,9,10)

InChI key

RKPHTRVPGYGVQD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Yu et al.
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases, 24(10), 608-610 (2002-01-05)
To observe and evaluate the effect and safety of pasinizid on treatment of the senile pulmonary tuberculosis with bacteriological positive. 115 cases of the senile pulmonary tuberculosis with bacteriological positive were divided into therapy group(group A) and control group(group B1
S S Zhang et al.
Journal of pharmaceutical and biomedical analysis, 17(4-5), 617-622 (1998-07-31)
Separation and determination of capreomycin (Cp), ofloxacin (Oflx) and pasiniazide (Ipa) in urine by high-performance capillary electrophoresis (HPCE) with 280 nm detection have been studied systematically. The calibration lines were linear in the range of 0.5 approximately 50 mg 1(-1)
Xian-min Zheng et al.
Di 1 jun yi da xue xue bao = Academic journal of the first medical college of PLA, 24(5), 574-575 (2004-05-21)
To observe the effects of the protocol combining levofloxacin, pasiniazide, M. Vaccae (V+D+M protocol) in the treatment of multi-drug resistant pulmonary tuberculosis (MDR-TB). Ninety-seven cases of MDR-TB randomized into V+D+M treatment protocol group (n=50) and control group (n=47) were observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service