Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2131

Sigma-Aldrich

Indulin B

Practical grade, powder

Synonym(s):

Acid Blue 20

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
50405
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Indulin B, practical grade

grade

practical grade

Quality Level

form

powder

solubility

water: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service