Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8760

Supelco

Hydroflumethiazide

analytical standard, ~97%

Synonym(s):

3,4-Dihydro-6-(trifluoromethyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F3N3O4S2
CAS Number:
Molecular Weight:
331.29
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

assay

~97%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O=S1(C2=CC(S(=O)(N)=O)=C(C(F)(F)F)C=C2NCN1)=O

InChI

1S/C8H8F3N3O4S2/c9-8(10,11)4-1-5-7(2-6(4)19(12,15)16)20(17,18)14-3-13-5/h1-2,13-14H,3H2,(H2,12,15,16)

InChI key

DMDGGSIALPNSEE-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

Hydroflumethiazide is a thiazide diuretic drug, which acts like a potent antihypertensive agent.[1] It is used in combination with spironolactone to minimize the excessive loss of potassium in hypokalemic patients who require diuretic therapy.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Hydroflumethiazide may be used as a reference standard in the determination of hydroflumethiazide in urine samples using micellar liquid chromatography coupled with fluorimetry detector (MLC).[3]

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hatch et al.
Clinical science (London, England : 1979), 86(3), 353-357 (1994-03-01)
1. The purpose of the present study was to examine the effects of various diuretics on intestinal oxalate transport. Transmural oxalate fluxes were measured across isolated, short-circuited tissue segments removed from rabbits and placed in Ussing chambers. 2. The net
S Suzuki et al.
No to shinkei = Brain and nerve, 38(2), 155-161 (1986-02-01)
The susceptibility to cerebral ischemia was studied in stroke-resistant spontaneously hypertensive rats (SHRSR) treated by a long-term antihypertensive treatment, and compared with untreated SHRSR and Wistar rats (WR). Male SHRSR, aged 8 weeks, were divided into two groups and a
D Hillaire-Buys et al.
Diabetologia, 27(6), 583-586 (1984-12-01)
LN 5330 is a new benzothiadiazine which is a structural analogue of diazoxide. Its effects in vivo were studied on blood glucose levels and insulin, glucagon and somatostatin secretion in normal dogs, and in vitro on glucagon and insulin secretion
E Selvaag
Photodermatology, photoimmunology & photomedicine, 12(6), 227-232 (1996-12-01)
A number of sulphonamide-derived oral antidiabetics and diuretics were investigated for phototoxic properties, using different sources of light, by means of a photohemolysis test. Photohemolysis was obtained after irradiation with UVA, visible light and solar simulating irradiation. No phototoxic properties
A A Awotedu
West African journal of medicine, 8(1), 89-90 (1989-01-01)
Haemospermia is an uncommon disorder of diverse aetiology, which includes diseases of the urethra, prostate and hypertension. There is no report of haemospermia in this environment and this communication describes a case associated with severe hypertension.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service