Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5395

Supelco

Hydrocortisone 17-valerate

analytical standard

Synonym(s):

11β,17,21-Trihydroxypregn-4-ene-3,20-dione 17-valerate, Cortisol 17-valerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H38O6
CAS Number:
Molecular Weight:
446.58
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(OC(=O)CCCC)C(=O)CO

InChI

1S/C26H38O6/c1-4-5-6-22(31)32-26(21(30)15-27)12-10-19-18-8-7-16-13-17(28)9-11-24(16,2)23(18)20(29)14-25(19,26)3/h13,18-20,23,27,29H,4-12,14-15H2,1-3H3/t18-,19-,20-,23+,24-,25-,26-/m0/s1

InChI key

FZCHYNWYXKICIO-FZNHGJLXSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erythematous patches and plaques on the chest with induration of the breasts. Metastatic papillary RCC in dermal lymphatics.
Yevgeniy Balagula et al.
Archives of dermatology, 147(10), 1215-1220 (2011-10-19)
K M Tramposch
Inflammation, 17(5), 531-536 (1993-10-01)
Synthetic glucocorticosteroids are widely used in clinical dermatology in the treatment of a number of inflammatory skin disorders. However, cutaneous side effects such as induction of skin atrophy are among the factors that limit the chronic use of this class
'Eye-catching' dermatitis for the skin surgeon.
R K Roenigk
Ophthalmic plastic and reconstructive surgery, 15(3), 149-150 (1999-06-04)
J Sefton et al.
Cutis, 32(1), 89-91 (1983-07-01)
Evaluations of the comparative efficacy and safety of a newly developed emollient ointment formulation of hydrocortisone valerate 0.2 percent were made in two double-blind, multicenter trials involving 145 patients with atopic dermatitis. Data are presented which indicate that hydrocortisone valerate
J G Alford et al.
Agents and actions, 37(3-4), 260-267 (1992-11-01)
We have previously shown that multiple topical applications, over 11 days, of the phorbol ester 12-O-tetradecanoylphorbol-13-acetate (TPA) induces a persistent inflammatory reaction characterized by edema, cell infiltration and epidermal hyperplasia. In order to characterize the cell infiltrate during the establishment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service