Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H167

Supelco

p-Hydroxybenzoylecgonine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO5
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccc(O)cc3)C(O)=O

InChI

1S/C16H19NO5/c1-17-10-4-7-12(17)14(15(19)20)13(8-10)22-16(21)9-2-5-11(18)6-3-9/h2-3,5-6,10,12-14,18H,4,7-8H2,1H3,(H,19,20)/t10-,12+,13-,14+/m0/s1

InChI key

GFOOTRIURAVHGP-AHLTXXRQSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service