Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

H1005

Hexobarbital

analytical standard

Synonym(s):

5-(1-Cyclohexenyl)-1,5-dimethylbarbituric acid, Evipal, Hexobarbitone, Methylhexabital

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27
UNSPSC Code:
41116107
PubChem Substance ID:
EC Number:
200-264-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

drug control

USDEA Schedule III; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN1C(=O)NC(=O)C(C)(C1=O)C2=CCCCC2

InChI

1S/C12H16N2O3/c1-12(8-6-4-3-5-7-8)9(15)13-11(17)14(2)10(12)16/h6H,3-5,7H2,1-2H3,(H,13,15,17)

InChI key

UYXAWHWODHRRMR-UHFFFAOYSA-N

General description

Hexobarbital is a widely used drug to analyze the capacity and ability of the liver to metabolize drugs.

Application

Hexobarbital may be used as a reference standard for the determination of hexobarbital and its metabolites, 3′-hydroxyhexobarbital and 3′-keto-hexobarbital from the biological samples by high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Intravenous anesthetic

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of hexobarbital and its main metabolites in mice by high-performance liquid chromatography.
Bocker R.
Journal of Chromatography. B, Biomedical Sciences and Applications, 228, 350-354 (1982)
Pharmacokinetics of hexobarbital in young and old rats.
Boonstra-Nieveld I H J, et al.
Mechanisms of Ageing and Development, 50(3), 289-298 (1989)
Martin Steinhaus et al.
Journal of agricultural and food chemistry, 57(7), 2882-2888 (2009-03-04)
Seventeen aroma-active volatiles, previously identified with high flavor dilution factors in fresh, pink Colombian guavas (Psidium guajava L.), were quantified by stable isotope dilution assays. On the basis of the quantitative data and odor thresholds in water, odor activity values
C D Sreedevi et al.
Journal of ethnopharmacology, 124(2), 171-175 (2009-05-09)
Sida acuta Burm. f. (Malvaceae) is used in Indian traditional medicine to treat liver disorders and is useful in treating nervous and urinary diseases and also disorders of the blood and bile. Evaluation of the hepatoprotective properties of the methanolic
Ozgür Devrim Can et al.
Phytotherapy research : PTR, 26(11), 1695-1700 (2012-03-08)
The present study was undertaken to investigate the putative activity of a methanol extract of Hypericum montbretti (Guttiferae) on the central nervous system. Rutin (1519 ppm) and quercitrin (784 ppm) were identified as the major phenolic compounds in the extract. When administered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service