Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0920500

Human coagulation factor IX concentrate BRP

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Factor IX human, Christmas factor, Plasma thromboplastin component

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

application(s)

pharmaceutical (small molecule)

format

neat

Storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Human coagulation factor IX concentrate BRP EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry Powell et al.
British journal of haematology, 168(1), 113-123 (2014-09-12)
In the phase 3 B-LONG [Recombinant Factor IX Fc Fusion Protein (rFIXFc) in Subjects with Haemophilia B] study, rFIXFc dosed every 1-2 weeks was safe and efficacious in previously treated subjects with haemophilia B. To date, there are no evaluations of
Jerry S Powell et al.
British journal of haematology, 168(1), 124-134 (2014-09-12)
In the phase 3 B-LONG (Recombinant Factor IX Fc Fusion Protein [rFIXFc] in Subjects With Haemophilia B) study, rFIXFc demonstrated a prolonged half-life compared with recombinant factor IX (rFIX), and safety and efficacy for prophylaxis and treatment of bleeding in
Amit C Nathwani et al.
The New England journal of medicine, 371(21), 1994-2004 (2014-11-20)
In patients with severe hemophilia B, gene therapy that is mediated by a novel self-complementary adeno-associated virus serotype 8 (AAV8) vector has been shown to raise factor IX levels for periods of up to 16 months. We wanted to determine
Massimo Franchini et al.
British journal of haematology, 165(5), 600-608 (2014-03-19)
Rituximab is a humanized chimeric anti-CD20 monoclonal antibody initially developed for the treatment of some haematological malignancies. Thanks to its ability to rapidly and specifically deplete B cells, it has also been used in a variety of autoimmune disorders, haematological
Robert F Sidonio et al.
American journal of hematology, 89(8), 831-836 (2014-05-20)
Little is known about rates of joint bleeding among females with FVIII/FIX deficiency or hemophilia carriers. In a cross-sectional study, we tested the hypothesis that females with FVIII or FIX deficiency enrolled in the Universal Data Collection (UDC) project had

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service