Key Documents
GXKT1013Q
Supelco
IsoMist Kit with Quartz Spray Chamber
for PerkinElmer Elan
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
KT-1013Q
compatibility
PerkinElmer Elan 500, 5000, 6000, 9000
PerkinElmer Elan 6100DRC, DRC II, DRC-e
Legal Information
IsoMist is a trademark of Glass Expansion Pty Ltd
Choose from one of the most recent versions:
Certificates of Analysis (COA)