Key Documents
GXKT1010
Supelco
IsoMist Kit
IsoMist Programmable Temperature Spray Chamber
About This Item
UNSPSC Code:
23151817
NACRES:
SB.32
quality
IsoMist Programmable Temperature Spray Chamber
manufacturer/tradename
(KT-1010)
compatibility
Agilent/HP 7500
Agilent/HP 7700
General description
Includes:
  • 20-809-3204 (IsoMist Module LOL with MB13 Spray Chamber V2)
  • 21-809-2881 (IsoMist Adaptor for Agilent 7500)
  • 21-809-2891 (PuraFlex Transfer Tube FB13-190 )
  • JC-13 (Metal Ball Joint Clip)
  • 21-809-2888 (IsoMist Alignment Tool for Agilent 7500)
Legal Information
IsoMist is a trademark of Glass Expansion Pty Ltd
Choose from one of the most recent versions:
Certificates of Analysis (COA)