GXAR301FSS2E
SeaSpray™ Nebulizer
2 mL/min with EzyFit & EzyLok
About This Item
quality
2 mL/min with EzyFit & EzyLok
manufacturer/tradename
(AR30-1-FSS2E)
compatibility
Agilent/Varian Liberty Series II Axial
Analytik Jena/Bruker, Varian aurora M90, 800-MS Series, ICP-MS
Leeman/Teledyne Leeman Series Axial/DV, Prodigy, Profile
Leeman/Teledyne Leeman Series Radial, Prodigy, Profile
PerkinElmer Elan 500, 5000, 6000, 9000
PerkinElmer Elan 6100DRC, DRC II, DRC-e
PerkinElmer Optima 3000RL, 3100RL, 3200RL
SII NanoTechlology/Seiko SPS/SPQ Series
Looking for similar products? Visit Product Comparison Guide
General description
- Includes EZT-075 EzyFit Connector with 0.75mm ID x 1.3mm OD x 700mm L tubing (1 unit)
- Includes EL-1 EzyLok Connector Kit
- This nebulizer supersedes AR30-1-FSS2
- Suggested Gas Flow 1 L/min
- Suggested Sample Uptake 2 mL/min
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service