Key Documents
GX318070060
Supelco
Tapered Alumina Injector
2.0 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
31-807-0060
compatibility
PerkinElmer Elan 500, 5000, 6000, 9000
PerkinElmer Elan 6100DRC, DRC II, DRC-e
PerkinElmer NexION 300
PerkinElmer Optima 3000RL, 3100RL, 3200RL
Choose from one of the most recent versions:
Certificates of Analysis (COA)