Key Documents
GX308083467
Supelco
D-Torch HF
without injector
About This Item
UNSPSC Code:
41115710
NACRES:
SB.32
quality
for Agilent 7700/7800/7900/8800
without injector
manufacturer/tradename
(30-808-3467)
compatibility
Agilent/HP 7700/7800/7900/8800
General description
Includes:
  • 31-808-3196 (Base and Inner Tube for D-Torch)
  • 31-808-3022 (Quartz Outer Tube for D-Torch)
  • 31-808-3072 (Injector Adaptor for Agilent 7500/7700 D-Torch)
  • 31-808-2962 (Injector Ferrule for D-Torch)
  • 31-808-3281 (PFA Connector with Dilution Port for Agilent 7700/7800/7900/8800 D-Torch)
  • SAFEGLOVE-med (Safety Gloves, Medium)
Choose from one of the most recent versions:
Certificates of Analysis (COA)