Key Documents
GX308083305
Supelco
D-Torch
without injector
Synonym(s):
Demountable Torch
About This Item
UNSPSC Code:
41115711
quality
for Agilent 7700/7900/8800
without injector
manufacturer/tradename
(30-808-3305)
compatibility
Agilent/HP 7700/7900/8800
General description
Includes:
  • 31-808-3196 (Base and Inner Tube for D-Torch)
  • 31-808-3022 (Quartz Outer Tube for D-Torch)
  • 31-808-3072 (Injector Adaptor for Agilent 7500/7700 D-Torch)
  • 31-808-2962 (Injector Ferrule for D-Torch)
  • 31-808-3303 (Quartz Connector for Agilent 7700/7800/7900/8800 D-Torch)
  • SAFEGLOVE-med (Safety Gloves, Medium)
Choose from one of the most recent versions:
Certificates of Analysis (COA)