Key Documents
GX308082937
Supelco
D-Torch
with Ceramic Outer Tube
Synonym(s):
Demountable Torch
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
quality
for Elan/NexION
with Ceramic Outer Tube
manufacturer/tradename
(30-808-2937)
compatibility
PerkinElmer 500, 5000, 6000, 9000
PerkinElmer Elan 6100DRC, DRC II, DRC-e
PerkinElmer NexION 300
General description
Includes:
  • 31-808-2847 (Base and Inner Tube for D-Torch)
  • 31-808-2849 (Ceramic Outer Tube for D-Torch)
  • 31-808-2818 (Retaining Ring for D-Torch Outer Tube)
  • SAFEGLOVE-med (Safety Gloves, Medium)
Choose from one of the most recent versions:
Certificates of Analysis (COA)