Key Documents
GX308081214
Supelco
Quartz Torch
1.8 mm injector with gas fittings
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
quality
1.8 mm injector with gas fittings
for Spectroflame
manufacturer/tradename
(30-808-1214)
compatibility
Spectro/Ametek SOP (Modula, CIROS, Genesis, ARCOS)
General description
Includes:
  • 30-808-0240 (Quartz Torch 1.8mm injector, 4mm OD side arms for Spectroflame)
  • 31-808-1166 (Gas Connector for Spectro Torch (Female))
  • 31-808-1167 (Gas Connector for Spectro Torch (Male))
Choose from one of the most recent versions:
Certificates of Analysis (COA)