Key Documents
GX308081212
Supelco
Quartz Torch
2.5 mm injector with gas fittings
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
quality
2.5 mm injector with gas fittings
for Spectro EOP
manufacturer/tradename
(30-808-1212)
compatibility
Spectro/Ametek EOP (Modula, CIROS, Genesis, ARCOS)
General description
Includes:
  • 30-808-0317 (Quartz Torch 2.5mm injector, 4mm OD side arms for Spectro EOP)
  • 31-808-1166 (Gas Connector for Spectro Torch (Female))
  • 31-808-1167 (Gas Connector for Spectro Torch (Male))
Choose from one of the most recent versions:
Certificates of Analysis (COA)