Key Documents
GX308080240
Supelco
Quartz Torch
1.8 mm injector, 4 mm OD side arms
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
quality
1.8 mm injector, 4 mm OD side arms
for Spectroflame
manufacturer/tradename
(30-808-0240)
compatibility
Spectro/Ametek SOP (Modula, CIROS, Genesis, ARCOS)
General description
Choose GAZ-04 GazFit Connectors for easy connection of gas tubes to torch side arms.
Choose from one of the most recent versions:
Certificates of Analysis (COA)