G9664
FreAlagin™ R gelatin
lyophilized powder
Synonym(s):
Gelatin from porcine skin
About This Item
biological source
Porcine skin
sterility
aseptically processed
form
lyophilized powder
mol wt
200-10,000
technique(s)
ELISA: suitable
cell culture | mammalian: suitable
immunocytochemistry: suitable
western blot: suitable
impurities
endotoxin, tested
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Components
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service