Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7020

Supelco

Glafenine hydrochloride

analytical standard

Synonym(s):

1-Glyceryl N-(7-chloro-4-quinolyl)anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN2O4 · HCl
CAS Number:
Molecular Weight:
409.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.OCC(O)COC(=O)c1ccccc1Nc2ccnc3cc(Cl)ccc23

InChI

1S/C19H17ClN2O4.ClH/c20-12-5-6-14-17(7-8-21-18(14)9-12)22-16-4-2-1-3-15(16)19(25)26-11-13(24)10-23;/h1-9,13,23-24H,10-11H2,(H,21,22);1H

InChI key

CEUMONXVSJOJIH-UHFFFAOYSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Zhang et al.
Molecular genetics & genomic medicine, 8(9), e1403-e1403 (2020-07-16)
Neural tube defects (NTDs) are severe congenital malformations. Diabetes during pregnancy is a risk factor for NTDs, but its mechanism remains elusive. Emerging evidence suggests that protein malonylation is involved in diabetes. Here, we report the correlation between histone lysine
Leanna Cheung et al.
Biochemical pharmacology, 93(3), 380-388 (2014-12-03)
Multidrug resistance protein 4 (MRP4) effluxes a wide variety of drugs and endogenous signaling molecules from cells and has been proposed as an attractive therapeutic target in several solid tumors, including neuroblastoma and colorectal cancer. MRP4 also regulates the pharmacokinetics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service