Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEF213

Sulfur in petrol

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

agency

ERM®

manufacturer/tradename

BAM

application(s)

pharmaceutical (small molecule)

format

matrix material

Analysis Note

For more information and to access certificate and report please visit the BAM webpage

Legal Information

ERM is a registered trademark of European Commission

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT RE 2

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-40.0 °F

flash_point_c

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service