Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMED102

Boron carbide

ERM®, certified reference material, powder

Synonym(s):

Carbon tetraboride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CB4
CAS Number:
Molecular Weight:
55.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

agency

ERM®

manufacturer/tradename

BAM

density

2.51 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

matrix material

SMILES string

B1=BB=C=B1

InChI

1S/CB4/c1-2-4-5-3-1

InChI key

NOJMLSPGQSYAIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information and to access certificate and report please visit the BAM webpage

Legal Information

ERM is a registered trademark of European Commission

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service