Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEC680K

Polyethylene (low level element content)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

agency

ERM®

application(s)

environmental
pharmaceutical (small molecule)

format

matrix material

Analysis Note

Legal Information

ERM is a registered trademark of European Commission

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

281.5 °F - closed cup

flash_point_c

138.6 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Schwabegger et al.
Synthetic metals, 161-66(19-20), 2058-2062 (2011-11-04)
We report on C(60) based organic field effect transistors (OFETs) that are well optimized for low voltage operation. By replacing commonly used dielectric layers by thin parylene films or by utilizing different organic materials like divinyltetramethyldisiloxane-bis(benzocyclo-butene) (BCB), low density polyethylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service