Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEB378

Copper alloy

ERM®, certified reference material, CuSn6, disc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

agency

ERM®

manufacturer/tradename

BAM

application(s)

pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information and to access certificate and report please visit the BAM webpage

Legal Information

ERM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service