Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEB325

Unalloyed zinc (trace elements)

ERM®, certified reference material

Synonym(s):

Zinc, unalloyed

Sign Into View Organizational & Contract Pricing

Select a Size

0.7 KG
$445.00

$445.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.7 KG
$445.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$445.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

format

matrix material

Analysis Note

For more information please see:
ERMEB325

Legal Information

ERM is a registered trademark of European Commission

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service