Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMCZ100

Fine dust (PM10-like) (PAH′s)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

storage temp.

2-8°C

Application

Fine dust (PM10-like) (PAH′s) has been used as a reference material in the determination of polycyclic aromatic hydrocarbons in air samples using high performance liquid chromatography (HPLC).[1]

Analysis Note

For more information please see:
ERMCZ100

Legal Information

ERM is a registered trademark of European Commission

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seasonal trends and spatial variations of PM10-bounded polycyclic aromatic hydrocarbons in Veneto Region, Northeast Italy
Masiol Masiol, et al.
Atmospheric Environment, 79, 811-821 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service