Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMCE464

Tuna fish (total Hg and methylmercury)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

agency

ERM®
certified by the BSC

composition

CH3Hg+, 5.50 mg/kg ±0.17 mg/kg by
total Hg, 5.24 mg/kg ±0.10 mg/kg by

manufacturer/tradename

JRC

matrix

fish

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Packaging

Glass bottle containing ~15 g powder

Analysis Note

For more information please see:
ERMCE464

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements, Metallorganics: Total Mercury (Hg), Methylmercury

Matrix Group: Fish and Seafood

Legal Information

ERM is a registered trademark of European Commission

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Kavčič et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 135, 110939-110939 (2019-11-08)
The effects of foliar Se biofortification (Se+) of the lettuce on the transfer and toxicity of Hg from soil contaminated with HgCl2 (H) and soil collected near the former Hg smelter in Idrija (I), to terrestrial food chain are explored

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service