Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMBF412E

Maize GMO Standard ERM-BF412

ERM®, certified reference material, 2% Bt-11, for the detection of GMO maize (Br-11) with PCR

Synonym(s):

GMO Standard ERM-BF412, Maize Bt-11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

agency

ERM®

quality

for the detection of GMO maize (Br-11) with PCR

composition

genetically modified Bt-11 maize, 2%

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBF412E

Legal Information

ERM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service