Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMBD283

Whole milk powder

ERM®, certified reference material, aflatoxin M1, low level

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701

grade

certified reference material

agency

ERM®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBD283

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Biotoxins / Mycotoxins: Aflatoxin M1

Matrix Group: Dairy products and eggs

Legal Information

ERM is a registered trademark of European Commission

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service