Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

E008054

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mp

58 °C (lit.)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

target_organs

Brain

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong-Uk Jin et al.
Magnetic resonance imaging, 31(7), 1143-1149 (2013-05-11)
The cochlear plays a vital role in the sense and sensitivity of hearing; however, there is currently a lack of knowledge regarding the relationships between mechanical transduction of sound at different intensities and frequencies in the cochlear and the neurochemical
Lisha Luo et al.
NMR in biomedicine, 25(12), 1360-1368 (2012-05-11)
The aim of this study was to provide data on the dose dependence of manganese-enhanced MRI (MEMRI) in the visual pathway of experimental rats and to study the toxicity of MnCl₂ to the retina. Sprague-Dawley rats were intravitreally injected with
Anne Bertrand et al.
NeuroImage, 64, 693-702 (2012-09-11)
The impairment of axonal transport by overexpression or hyperphosphorylation of tau is well documented for in vitro conditions; however, only a few studies on this phenomenon have been conducted in vivo, using invasive procedures, and with contradictory results. Here we
Geoffrey J Topping et al.
Medical physics, 40(4), 042502-042502 (2013-04-06)
Manganese(II) is employed as a contrast agent with magnetic resonance imaging (MRI) for study of neuronal activation in rats and mice. However, at the concentrations required for MRI, Mn may induce pharmacological or toxic effects. Positron emission tomography (PET) imaging
A G Kanthasamy et al.
Toxicology letters, 214(3), 288-295 (2012-09-22)
The role of normal cellular prion protein (PrP) remains to be fully elucidated; however, the protein is crucial for the infection and progression of prion diseases. Recent evidence indicates that PrP is a metalloprotein since the octapeptide repeat sequences in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service