Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW456217

Sigma-Aldrich

Corning® Supersomes SupersomesEnzyme CYP2D6′1+OR Human 0.5mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−70°C

Application

Drug metabolism and ADME/Tox studies

Legal Information

Corning is a registered trademark of Corning, Inc.
Supersomes is a trademark of Corning, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service