Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW454560

Sigma-Aldrich

Corning® Gentest Medium HepatoCryo Recovery Hi Viability

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

shipped in

wet ice

storage temp.

2-8°C

General description

The use of this product can enhance post-thaw cell viability for cryopreserved primary hepatocytes. Only a single centrifugation step is needed when using this product.

Features and Benefits

Features and Benefits
  • Improves post-thaw cell viability and yield for cryopreserved primary hepatocytes.
  • Only a single centrifugation is needed, which saves time for researchers.
  • Each recovery medium can be used with up to five vials of cryopreserved.
  • Non-cytotoxic nature and suitable to use with all cryopreserved hepatocytes in assays requiring plated cells, such as induction, transporter, metabolism, and toxicity studies.
  • The recovery medium is also available together with a high efficiency plating medium in the Corning Gentest high viability recovery kit (Cat No 454534).

Legal Information

Corning is a registered trademark of Corning, Inc.
Gentest is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service