Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW452501

Sigma-Aldrich

Corning® Gentest Microsome Rat Liver Pool Male 0.5 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−70°C

General description

Corning® Gentest Microsome Rat Liver Pool Male 0.5 mL is prepared from sexually mature male Sprague-Dawley rat livers. It is characterized for relevant cytochrome P450 enzyme activities.

application

Drug transporters and ADME/Tox studies

Features and Benefits

  • Poole liver microtomes prepared from sexually mature male Sprague-Dawley rats
  • Large lot size available
  • High manufacturing standards to reduce lot-to-lot variability and to ensure reproducibility
  • Available size: 0.5 mL
  • Protein concentration: 20 mg/mL in 250mM sucrose
  • Shelf life: this product is stable for more than ten years when stored properly

Legal Information

Corning is a registered trademark of Corning, Inc.
Gentest is a trademark of Corning, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service