Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW451710

Sigma-Aldrich

Corning® Gentest Bupropion HCL 25 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

dry ice

Storage temp.

−20°C

Legal Information

Corning is a registered trademark of Corning, Inc.
Gentest is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service