Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW451201

Sigma-Aldrich

Corning® Gentest Buffer Phosphate 0.5M 500 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

concentration

0.5 M

shipped in

ambient

storage temp.

room temp

General description

Corning® Gentest Phosphate Buffer with pH 7, is used as assay buffer for cytochrome P450 and Phase II enzyme drug metabolism assays.

Application

Corning® Gentest Buffer Phosphate 0.5M 500 mL may be used as a component of homogenizing buffer for isolating microsomes from liver tissues.[1][2] It has also been used as a buffer component in in vitro assay with human liver microsomes.[3]

Legal Information

Corning is a registered trademark of Corning, Inc.
Gentest is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Els Van Peer et al.
Basic & clinical pharmacology & toxicology, 117(5), 350-357 (2015-04-22)
In view of paediatric drug development, regulatory authorities often request safety studies in juvenile animals, including minipigs. Unfortunately, knowledge on the ontogeny of the biotransformation processes in animal models remains scarce and impedes a correct interpretation of the toxicity findings.
In vivo and in vitro metabolism of the novel synthetic cannabinoid 5F-APINAC
Appolonova SA, et al.
Forensic Toxicology, 160-171 (2020)
Els Van Peer et al.
Pharmaceutical research, 34(4), 750-764 (2017-01-18)
In view of pediatric drug development, juvenile animal studies are gaining importance. However, data on drug metabolizing capacities of juvenile animals are scarce, especially in non-rodent species. Therefore, we aimed to characterize the in vitro biotransformation of four human CYP450

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service