Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW356778

Corning® PureCoat Flask T75 Carboxyl 5pac 50cas

Synonym(s):

Corning container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
Pricing and availability is not currently available.

shipped in

wet ice

storage temp.

room temp

Legal Information

Corning is a registered trademark of Corning, Inc.
PureCoat is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service