DLW356650
Corning® BioCoat™ Plate 96w Col I Wht/Clr 5pac 50cas
Synonym(s):
plastic coated cell culture plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
size:
6 wells
size
6 wells
suitability
suitable for (cell culture)
shipped in
wet ice
storage temp.
room temp
Legal Information
BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service