Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW356236

Sigma-Aldrich

Corning® Collagen I Rat Tail 1 (10x100 mg)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

biological source

rat tail

assay

>90% (SDS-PAGE)

form

liquid (in 0.02 N acetic acid)

packaging

pkg of 1 g

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

Corning® collagen I, rat tail, 1g (10 x 100 mg), is used either as a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function.

Other Notes

Source:
Rat Tail Tendon

Quality:
  • Purity = 90% by SDS-PAGE
  • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested for formation of a firm gel up to a dilution of 1:10; further dilution will decrease the rigidity of the gel
  • Found negative for bacteria, fungi, and mycoplasma



Formulation
Liquid in 0.02 N acetic acid, concentration range 3-4 mg/mL

Preparation and Storage
Stable for at least three months at 2° to 8°C. Do not freeze.

Handling
Effective as a gel or thin coating at a recommended concentration of 5-10 μg/cm2 of growth surface depending on cell type.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service