Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW356231

Sigma-Aldrich

Corning® Matrigel® Matrigel Matrix GFR PhenolRF Mouse 10 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

shipped in

dry ice

Storage temp.

−20°C

General description

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Quality

Quality
  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing.
  • Extensive PCR testing is performed to screen for a number of pathogens.
  • Tested and found negative for bacteria, fungi, and mycoplasma.
  • Protein concentrations determined by Lowry method.
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay.
  • Matrigel Matrix gel stability is tested for a period of 14 days at 37°C.
  • Biological activity is determined for each lot using a neurite outgrowth assay.

Other Notes

Source:
Engelbreth-Holm-Swarm Mouse Tumor.

Formulation:
High concentration laminin/entactin complex frozen in Dulbecco′s PBS.

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammedin Deliorman et al.
Bio-protocol, 10(18), e3764-e3764 (2021-03-05)
This protocol describes a simple method to cryopreserve mammalian cells within filter papers as an alternative to conventional slow-freezing approach. The method involves treating paper fibers with fibronectin, using low concentrations of the cryoprotectant dimethyl sulfoxide (DMSO), and slow freezing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service