Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW355058

SAFC

Corning® BioCoat Kit Media Intestinal Epithel Diff Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

shipped in

wet ice

storage temp.

2-8°C

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service