Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW355006

Sigma-Aldrich

Corning® Mito+ Serum Extender 5 L Equiv

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

shipped in

wet ice

storage temp.

2-8°C

General description

Corning® MITO+ Serum Extender, 5 mL, is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steroid hormones). 5 mL (5L equivalent), 1/pack, formerly Cat #50006

Storage and Stability

Shelf life: ≥3 months from date of shipment at 2-8°C (lyophilized) or ≥ three months at -20°C (solubilized)

Legal Information

Corning is a registered trademark of Corning, Inc.

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyen Katrina Kan et al.
Purinergic signalling, 16(3), 327-336 (2020-06-26)
Gliomas, the most common primary brain cancer, are highly infiltrative and extremely difficult to treat. Despite advancements, current treatment is limited, with patients surviving for a median of 14-15 months post-diagnosis. Previous research has demonstrated the upregulation of a purinergic receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service