Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354717

Corning® PureCoat Plate 96w Amine Blk/Clr 5pac 5cas

Synonym(s):

plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
size:
6 wells
Pricing and availability is not currently available.

size

6 wells

suitability

suitable for (cell culture)

shipped in

wet ice

storage temp.

room temp

Legal Information

Corning is a registered trademark of Corning, Inc.
PureCoat is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service